5-Methyl-3-isoxazolcarbonylchlorid
Lieferant: Thermo Scientific
CC10402.0001
CC10402DA
CC10402EA
MAYBCC10402.0001EA
0
EUR
MAYBCC10402.0001
MAYBCC10402DA
MAYBCC10402EA
5-Methyl-3-isoxazolcarbonylchlorid
5-Methyl-3-isoxazolcarbonylchlorid
Formel:
C₅H₄ClNO₂ Molecular Weight: 145,55 g/mol |
MDL:
MFCD00085127 CAS-Nummer: 39499-34-8 EINECS: 254-475-6 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...