3,5-Dimethyl-4-isoxazolboronsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (dimethyl-1,2-oxazol-4-yl)boronic acid

H26893.MD H26893.03 H26893.06
H26893.MDEA 0 EUR
H26893.MD H26893.03 H26893.06
3,5-Dimethyl-4-isoxazolboronsäure ≥97%
3,5-Dimethylisoxazole-4-boronic acid

Purity: 97% CAS No.: 16114-47-9 Molecular Formula: C5H8BNO3 Formula Weight: 140.93 Storage Temperature: 2°C to 8°C Melting Point: 106-108° MDL No.: MFCD02677945

Formel: C₅H₈BNO₃
Molecular Weight: 140,93 g/mol
Schmelzpunkt: 106…108 °C
MDL: MFCD02677945
CAS-Nummer: 16114-47-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR