3,5-Dimethyl-4-isoxazolboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (dimethyl-1,2-oxazol-4-yl)boronic acid

720410-250MG 720410-1G
SIAL720410-250MGEA 17.5 EUR
SIAL720410-250MG SIAL720410-1G
3,5-Dimethyl-4-isoxazolboronsäure, Sigma-Aldrich®
3,5-Dimethylisoxazole-4-boronic acid
Formel: C₅H₈BNO₃
Molecular Weight: 140,93 g/mol
MDL: MFCD02677945
CAS-Nummer: 16114-47-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR