3,5-Dimethyl-4-isoxazolcarbonylchlorid

Lieferant: FLUOROCHEM

CFF270.5 CFF270.10 CFF270.25 017317-10G 017317-25G 017317-1G 017317-5G
CHMPCFF270.5EA 666 EUR
CHMPCFF270.5 CHMPCFF270.10 CHMPCFF270.25 FLUO017317-10G FLUO017317-25G FLUO017317-1G FLUO017317-5G
3,5-Dimethyl-4-isoxazolcarbonylchlorid
3,5-Dimethyl-4-isoxazolcarbonylchlorid
Formel: C₆H₆ClNO₂
MDL: MFCD00052555
CAS-Nummer: 31301-45-8
UN: 3265
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR