3,5-Dimethyl-4-isoxazolcarbonylchlorid

Lieferant: Thermo Scientific

CC00602.0001 CC00602.250 CC00602DA
MAYBCC00602.0001EA 0 EUR
MAYBCC00602.0001 MAYBCC00602.250 MAYBCC00602DA
3,5-Dimethyl-4-isoxazolcarbonylchlorid
3,5-Dimethyl-4-isoxazolcarbonylchlorid
Formel: C₆H₆ClNO₂
Molecular Weight: 159,57 g/mol
MDL: MFCD00052555
CAS-Nummer: 31301-45-8
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR