3,5-Dimethyl-4-isoxazolcarbonsäure ≥99%

Lieferant: Thermo Scientific

Synonyme: 3,5-Dimethylisoxasole-4-carboxylic acid

A15419.06 A15419.14
A15419.06EA 107 EUR
A15419.06 A15419.14
3,5-Dimethyl-4-isoxazolcarbonsäure ≥99%
3,5-Dimethylisoxazole-4-carboxylic acid

Purity: 99% CAS No.: 2510-36-3 Molecular Formula: C6H7NO3 Formula Weight: 141.13 Melting Point: 140-142° Beilstein Reference: 115819

Formel: C₆H₇NO₃
Molecular Weight: 141,13 g/mol
Schmelzpunkt: 140…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051657
CAS-Nummer: 2510-36-3
EINECS: 219-724-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR