3,5-Dimethyl-4-isoxazolcarbonsäure

Lieferant: BIONET RESEARCH

Synonyme: 3,5-Dimethylisoxasole-4-carboxylic acid

11X-0244.1 11X-0244.10 11X-0244.5 11X-0244.500
BIOT11X-0244.1EA 0 EUR
BIOT11X-0244.1 BIOT11X-0244.10 BIOT11X-0244.5 BIOT11X-0244.500
3,5-Dimethyl-4-isoxazolcarbonsäure
3,5-Dimethylisoxazole-4-carboxylic acid
Formel: C₆H₇NO₃
Molecular Weight: 141.13 g/mol
MDL: MFCD00051657
CAS-Nummer: 2510-36-3
EINECS: 219-724-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR