3,5-Dimethyl-4-isoxazolcarbonsäure
Lieferant: FLUOROCHEM
Synonyme:
3,5-Dimethylisoxasole-4-carboxylic acid
CFF176.5
CFF176.25
017249-5G
017249-25G
CHMPCFF176.5EA
481
EUR
CHMPCFF176.5
CHMPCFF176.25
FLUO017249-5G
FLUO017249-25G
3,5-Dimethyl-4-isoxazolcarbonsäure
3,5-Dimethylisoxazole-4-carboxylic acid
Formel:
C₆H₇NO₃ Molecular Weight: 141.13 g/mol |
MDL:
MFCD00051657 CAS-Nummer: 2510-36-3 EINECS: 219-724-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...