Fluoren-2-carbaldehyd ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: Fluorene-2-carboxaldehyde

A14911.06 A14911.14 A14911.22
A14911.06EA 77.4 EUR
A14911.06 A14911.14 A14911.22
Fluoren-2-carbaldehyd ≥99%
Fluoren-2-carbaldehyd

Purity: 99% CAS No.: 30084-90-3 Molecular Formula: C14H10O Formula Weight: 194.23 Melting Point: 82-86° Beilstein Reference: 2255852

Formel: C₁₄H₁₀O
Molecular Weight: 194.23 g/mol
Schmelzpunkt: 82…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001123
CAS-Nummer: 30084-90-3
EINECS: 250-035-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR