N-Docosanoyltaurin

Lieferant: Cayman Chemical

10007288-1 10007288-10 10007288-5
CAYM10007288-1EA 112 EUR
CAYM10007288-1 CAYM10007288-10 CAYM10007288-5
N-Docosanoyltaurin
N-Docosanoyltaurin
Formel: C₂₄H₄₉NO₄S
CAS-Nummer: 783284-48-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR