Glycinamid Hydrochlorid (H-Gly-NH2.HCl) ≥98%
Lieferant: Thermo Scientific
Synonyme: H-Gly-NH2.HCl
A11128.06 A11128.14 A11128.22
A11128.06EA 31.3 EUR
A11128.06 A11128.14 A11128.22
Glycinamid Hydrochlorid (H-Gly-NH2.HCl) ≥98%
Glycinamid Hydrochlorid (H-Gly-NH2.HCl)
Purity: 98% CAS No.: 1668-10-6 Molecular Formula: NH2C(O)CH2NH2.HCl Formula Weight: 110.54 Melting Point: ca 204° dec. Beilstein Reference: 3554199
Formel: C₂H₇ClN₂O
Molecular Weight: 110,54 g/mol
Schmelzpunkt: ∼ 204 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013008
CAS-Nummer: 1668-10-6
EINECS: 216-789-1