Glycinamid Hydrochlorid (H-Gly-NH2.HCl)

Lieferant: BACHEM BIOCHEMICA

Synonyme: H-Gly-NH2.HCl

4025766.0100 4025766.0500
BACE4025766.0100EA 0 EUR
BACE4025766.0100 BACE4025766.0500
Glycinamid Hydrochlorid (H-Gly-NH2.HCl)
Glycinamid Hydrochlorid (H-Gly-NH2.HCl)
Formel: C₂H₇ClN₂O
Molecular Weight: 110.54 g/mol
MDL: MFCD00013008
CAS-Nummer: 1668-10-6
EINECS: 216-789-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR