2-Bromisobuttersäureamid
Lieferant: COMBI-BLOCKS
Synonyme:
2-Bromo-2-methylpropanamide
CA-4521-25G
CA-4521-5G
CA-4521-1G
COBBCA-4521-25GEA
179
EUR
COBBCA-4521-25G
COBBCA-4521-5G
COBBCA-4521-1G
2-Bromisobuttersäureamid
2-Bromisobuttersäureamid
Formel:
C₄H₈BrNO Molecular Weight: 166.0164 g/mol |
MDL:
MFCD08234837 CAS-Nummer: 7462-74-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...