2-(3-Trifluormethyl-α-toluolsulfonyl)thioacetamid

Lieferant: FLUOROCHEM

008436-1G 008436-5G
FLUO008436-1GEA 0 EUR
FLUO008436-1G FLUO008436-5G
2-(3-Trifluormethyl-α-toluolsulfonyl)thioacetamid
2-(3-Trifluormethyl-α-toluolsulfonyl)thioacetamid
Formel: C₁₀H₁₀F₃NO₂S₂
MDL: MFCD00052940
CAS-Nummer: 175276-83-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR