Methyl-trans-4-oxo-2-pentenoat, Sigma-Aldrich®

Lieferant: Merck

75944-1G 75944-5G
SIAL75944-1GEA 243 EUR
SIAL75944-1G SIAL75944-5G
Methyl-trans-4-oxo-2-pentenoat, Sigma-Aldrich®
Methyl-trans-4-oxo-2-pentenoat
Formel: C₆H₈O₃
MDL: MFCD00059331
CAS-Nummer: 2833-24-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR