4-(4-Bromphenoxy)benzaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(4-Bromo-phenoxy)benzaldehyde

M-Clarity™ Qualitätsstufe = MQ100

661295-1G 661295-5G
SIAL661295-1GEA 69.6 EUR
SIAL661295-1G SIAL661295-5G
4-(4-Bromphenoxy)benzaldehyd, Sigma-Aldrich®
4-(4-Bromophenoxy)benzaldehyde
Formel: C₁₃H₉BrO₂
Molecular Weight: 277,12 g/mol
MDL: MFCD08276811
CAS-Nummer: 69240-56-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR