N-Methyl-2-brom-4-chloranilin Hydrochlorid
Lieferant: COMBI-BLOCKS
HC-2882-5G
HC-2882-1G
COBBHC-2882-5GEA
1090
EUR
COBBHC-2882-5G
COBBHC-2882-1G
N-Methyl-2-brom-4-chloranilin Hydrochlorid
N-Methyl-2-brom-4-chloranilin Hydrochlorid
Formel:
C₇H₈BrCl₂N |
MDL:
MFCD13195691 CAS-Nummer: 1199773-47-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...