N-Methyl-2-brom-5-chloranilin Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-2888-5G HC-2888-1G
COBBHC-2888-5GEA 1180 EUR
COBBHC-2888-5G COBBHC-2888-1G
N-Methyl-2-brom-5-chloranilin Hydrochlorid
N-Methyl-2-brom-5-chloranilin Hydrochlorid
Formel: C₇H₈BrCl₂N
MDL: MFCD11847062
CAS-Nummer: 1187386-26-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR