N-Methyl-3-bromanilin Hydrochlorid
Lieferant: COMBI-BLOCKS
Synonyme:
3-Bromo-N-methylaniline hydrochloride
HC-2799-5G
HC-2799-1G
COBBHC-2799-5GEA
121
EUR
COBBHC-2799-5G
COBBHC-2799-1G
N-Methyl-3-bromanilin Hydrochlorid
N-Methyl-3-bromanilin Hydrochlorid
Formel:
C₇H₉BrClN Molecular Weight: 222,5101 g/mol |
MDL:
MFCD12756434 CAS-Nummer: 152814-26-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...