2-(4-Methoxyphenoxy)benzaldehyd

Lieferant: BIONET RESEARCH

11N-047.500 11N-047.1
BIOT11N-047.500EA 0 EUR
BIOT11N-047.500 BIOT11N-047.1
2-(4-Methoxyphenoxy)benzaldehyd
2-(4-Methoxyphenoxy)benzaldehyde
Formel: C₁₄H₁₂O₃
Molecular Weight: 228.2400 g/mol
MDL: MFCD01568732
CAS-Nummer: 19434-36-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR