2-Amino-5-chlorbenzaldehyd 97%

Lieferant: Thermo Scientific
Achtung

L17515.03 L17515.06
L17515.03EA 61.8 EUR
L17515.03 L17515.06
2-Amino-5-chlorbenzaldehyd 97%
2-Amino-5-chlorobenzaldehyde

Purity: 97% CAS No.: 20028-53-9 Molecular Formula: C7H6ClNO Formula Weight: 155.58 Melting Point: 68-74° Beilstein Reference: 2716096

Formel: C₇H₆ClNO
Molecular Weight: 155,58 g/mol
Schmelzpunkt: 68…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093936
CAS-Nummer: 20028-53-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR