4-(2,4-Dichlorphenoxy)benzaldehyd

Lieferant: BIONET RESEARCH

12N-058.500 12N-058.1
BIOT12N-058.500EA 0 EUR
BIOT12N-058.500 BIOT12N-058.1
4-(2,4-Dichlorphenoxy)benzaldehyd
4-(2,4-Dichlorphenoxy)benzaldehyd
Formel: C₁₃H₈Cl₂O₂
MDL: MFCD01568767
CAS-Nummer: 86309-06-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR