2-(3-Chlorphenoxy)benzaldehyd

Lieferant: BIONET RESEARCH

12N-067.500 12N-067.1 12N-067.5
BIOT12N-067.500EA 0 EUR
BIOT12N-067.500 BIOT12N-067.1 BIOT12N-067.5
2-(3-Chlorphenoxy)benzaldehyd
2-(3-Chlorophenoxy)benzaldehyde
Formel: C₁₃H₉ClO₂
Molecular Weight: 232.6624 g/mol
MDL: MFCD01568769
CAS-Nummer: 181297-72-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR