4-Chlor-2-fluor-6-iodanilin 96%

Lieferant: Thermo Scientific
Gefahr

L17658.03 L17658.06
L17658.03EA 31.5 EUR
L17658.03 L17658.06
4-Chlor-2-fluor-6-iodanilin 96%
4-Chlor-2-fluor-6-iodanilin

Purity: 96% CAS No.: 216393-67-8 Molecular Formula: C6H4ClFIN Formula Weight: 271.46 Melting Point: 39-41° UN No.: 2811 MDL No.: MFCD01318122

Formel: C₆H₄ClFIN
Molecular Weight: 271,46 g/mol
Schmelzpunkt: 39…41 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01318122
CAS-Nummer: 216393-67-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR