N-Methyl-2-brom-5-fluoranilin Hydrochlorid
Lieferant: COMBI-BLOCKS
HC-2877-5G
HC-2877-1G
COBBHC-2877-5GEA
574
EUR
COBBHC-2877-5G
COBBHC-2877-1G
N-Methyl-2-brom-5-fluoranilin Hydrochlorid
N-Methyl-2-brom-5-fluoranilin Hydrochlorid
Formel:
C₇H₈BrClFN |
MDL:
MFCD13195688 CAS-Nummer: 1199773-27-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...