N-Methyl-3,4-dichloranilin Hydrochlorid
Lieferant: COMBI-BLOCKS
HC-2856-5G
HC-2856-25G
HC-2856-1G
COBBHC-2856-5GEA
0
EUR
COBBHC-2856-5G
COBBHC-2856-25G
COBBHC-2856-1G
N-Methyl-3,4-dichloranilin Hydrochlorid
N-Methyl-3,4-dichloranilin Hydrochlorid
Formel:
C₇H₈Cl₃N |
MDL:
MFCD12913955 CAS-Nummer: 1187385-65-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...