N-Methyl-3,5-dichloranilin Hydrochlorid

Lieferant: COMBI-BLOCKS

Synonyme: 3,5-Dichloro-N-methylaniline hydrochloride

HC-2901-1G HC-2901-5G
COBBHC-2901-1GEA 149 EUR
COBBHC-2901-1G COBBHC-2901-5G
N-Methyl-3,5-dichloranilin Hydrochlorid
N-Methyl-3,5-dichloranilin Hydrochlorid
Formel: C₇H₈Cl₃N
Molecular Weight: 212.504 g/mol
MDL: MFCD08456850
CAS-Nummer: 1197239-04-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR