2-(4-Fluorphenoxy)benzaldehyd

Lieferant: BIONET RESEARCH

12N-071.500 12N-071.1 12N-071.5 12N-071.10
BIOT12N-071.500EA 0 EUR
BIOT12N-071.500 BIOT12N-071.1 BIOT12N-071.5 BIOT12N-071.10
2-(4-Fluorphenoxy)benzaldehyd
2-(4-Fluorophenoxy)benzaldehyde
Formel: C₁₃H₉FO₂
Molecular Weight: 216.21 g/mol
MDL: MFCD01568770
CAS-Nummer: 320423-61-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR