4-(4-Fluorphenoxy)benzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

661287-1G 661287-5G
SIAL661287-1GEA 106 EUR
SIAL661287-1G SIAL661287-5G
4-(4-Fluorphenoxy)benzaldehyd, Sigma-Aldrich®
4-(4-Fluorophenoxy)benzaldehyde
Formel: C₁₃H₉FO₂
Molecular Weight: 216.21 g/mol
MDL: MFCD01631896
CAS-Nummer: 137736-06-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR