N-Methyl-2,4-difluoranilin Hydrochlorid
Lieferant: COMBI-BLOCKS
HC-2854-5G
HC-2854-1G
COBBHC-2854-5GEA
742
EUR
COBBHC-2854-5G
COBBHC-2854-1G
N-Methyl-2,4-difluoranilin Hydrochlorid
N-Methyl-2,4-difluoranilin Hydrochlorid
Formel:
C₇H₈ClF₂N |
MDL:
MFCD11110410 CAS-Nummer: 1187386-15-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...