Methyl-2-(brommethyl)-3-chlorbenzoat

Lieferant: CHEMPUR

03-0013.1 03-0013.5
CHMP03-0013.1EA 0 EUR
CHMP03-0013.1 CHMP03-0013.5
Methyl-2-(brommethyl)-3-chlorbenzoat
Methyl-2-(brommethyl)-3-chlorbenzoat
Formel: C₉H₈BrClO₂
MDL: MFCD04114322
CAS-Nummer: 188187-03-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR