Methyl-2-(brommethyl)-6-chlorbenzoat

Lieferant: CHEMPUR

03-0016.1 03-0016.5
CHMP03-0016.1EA 0 EUR
CHMP03-0016.1 CHMP03-0016.5
Methyl-2-(brommethyl)-6-chlorbenzoat
Methyl-2-(brommethyl)-6-chlorbenzoat
Formel: C₉H₈BrClO₂
MDL: MFCD04114325
CAS-Nummer: 482578-63-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR