Methyl-2-(brommethyl)-3-nitrobenzoat
Lieferant: CHEMPUR
Synonyme:
Methyl 2-(bromomethyl)-3-nitrobenzoate
03-0004.1
03-0004.5
CHMP03-0004.1EA
0
EUR
CHMP03-0004.1
CHMP03-0004.5
Methyl-2-(brommethyl)-3-nitrobenzoat
Methyl-2-(brommethyl)-3-nitrobenzoat
Formel:
C₉H₈BrNO₄ Molecular Weight: 274,07 g/mol |
MDL:
MFCD04114315 CAS-Nummer: 98475-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...