Methyl-2-(brommethyl)-5-nitrobenzoat

Lieferant: CHEMPUR

Synonyme: Methyl 2-(bromomethyl)-5-nitrobenzoate

03-0001.1 03-0001.5
CHMP03-0001.1EA 0 EUR
CHMP03-0001.1 CHMP03-0001.5
Methyl-2-(brommethyl)-5-nitrobenzoat
Methyl-2-(brommethyl)-5-nitrobenzoat
Formel: C₉H₈BrNO₄
Molecular Weight: 274,0681 g/mol
MDL: MFCD04114313
CAS-Nummer: 90725-68-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR