Methyl-2-(brommethyl)-6-nitrobenzoat

Lieferant: CHEMPUR

Synonyme: Methyl 2-(bromomethyl)-6-nitrobenzoate

03-0002.1 03-0002.5
CHMP03-0002.1EA 0 EUR
CHMP03-0002.1 CHMP03-0002.5
Methyl-2-(brommethyl)-6-nitrobenzoat
Methyl-2-(brommethyl)-6-nitrobenzoat
Formel: C₉H₈BrNO₄
Molecular Weight: 274,0700 g/mol
MDL: MFCD04114314
CAS-Nummer: 61940-21-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR