2'-Bromo-4'-(trifluoromethyl)acetanilide

Lieferant: COMBI-BLOCKS

Synonyme: 4-Acetamido-3-bromobenzotrifluoride, N-[2-Bromo-4-(trifluoromethyl)phenyl]acetamide

AN-1241-5G AN-1241-25G AN-1241-1G
COBBAN-1241-5GEA 137 EUR
COBBAN-1241-5G COBBAN-1241-25G COBBAN-1241-1G
2'-Bromo-4'-(trifluoromethyl)acetanilide
2'-Bromo-4'-(trifluoromethyl)acetanilide
Formel: C₉H₇BrF₃NO
Molecular Weight: 282.057 g/mol
MDL: MFCD00067782
CAS-Nummer: 175135-49-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR