Methyl-2-(brommethyl)-5-fluorbenzoat
Lieferant: CHEMPUR
03-0014.1
03-0014.5
CHMP03-0014.1EA
0
EUR
CHMP03-0014.1
CHMP03-0014.5
Methyl-2-(brommethyl)-5-fluorbenzoat
Methyl-2-(brommethyl)-5-fluorbenzoat
Formel:
C₉H₈BrFO₂ Molecular Weight: 247,06 g/mol |
MDL:
MFCD04114323 CAS-Nummer: 138786-65-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...