2,6-Dinitro-4-(trifluormethyl)phenol

Lieferant: FLUOROCHEM

010457-25G 010457-5G
FLUO010457-25GEA 425 EUR
FLUO010457-25G FLUO010457-5G
2,6-Dinitro-4-(trifluormethyl)phenol
2,6-Dinitro-4-(trifluormethyl)phenol
Formel: C₇H₃F₃N₂O₅
MDL: MFCD00194260
CAS-Nummer: 393-77-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR