4-(Trifluoromethyl)diphenylether

Lieferant: CHEMPUR

Synonyme: 1-Phenoxy-4-(trifluoromethyl)benzene

FL5856.1 FL5856.5 FL5856.10
CHMPFL5856.1EA 0 EUR
CHMPFL5856.1 CHMPFL5856.5 CHMPFL5856.10
4-(Trifluoromethyl)diphenylether
4-(Trifluoromethyl)diphenylether
Formel: C₁₃H₉F₃O
Molecular Weight: 238,21 g/mol
MDL: MFCD01631641
CAS-Nummer: 2367-02-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR