4-Toluolsulfonyl-(3-trifluormethylanilid)
Lieferant: MENAI
Synonyme:
4-Methyl-N-[3-(trifluoromethyl)phenyl]benzenesulphonamide
ST655.5
ST655.2
ST655.10
ST655.1
MENAST655.5EA
0
EUR
MENAST655.5
MENAST655.2
MENAST655.10
MENAST655.1
4-Toluolsulfonyl-(3-trifluormethylanilid)
4-Toluolsulfonyl-(3-trifluormethylanilid)
Formel:
C₁₄H₁₂F₃NO₂S |
MDL:
MFCD00245086 CAS-Nummer: 1584-58-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...