2-Morpholino-3-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-8925-1G PN-8925-250M
COBBPN-8925-1GEA 410 EUR
COBBPN-8925-1G COBBPN-8925-250M
2-Morpholino-3-pyridinboronsäurepinakolester
2-Morpholino-3-pyridinboronsäurepinakolester
Formel: C₁₅H₂₃BN₂O₃
MDL: MFCD11044431
CAS-Nummer: 1150561-72-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR