2-Morpholino-4-pyridinboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)morpholine
PN-9128-5G
PN-9128-1G
COBBPN-9128-5GEA
1490
EUR
COBBPN-9128-5G
COBBPN-9128-1G
2-Morpholino-4-pyridinboronsäurepinakolester
2-Morpholino-4-pyridinboronsäurepinakolester
Formel:
C₁₅H₂₃BN₂O₃ |
MDL:
MFCD11054039 CAS-Nummer: 888721-86-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...