2-Morpholino-3-pyridinboronsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

705039-1G 705039-250MG
SIAL705039-1GEA 329 EUR
SIAL705039-1G SIAL705039-250MG
2-Morpholino-3-pyridinboronsäure, Sigma-Aldrich®
2-Morpholino-3-pyridinboronsäure
Formel: C₉H₁₃BN₂O₃
MDL: MFCD11867876
CAS-Nummer: 1218790-86-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR