2-Morpholino-5-pyridinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-(Morpholino)pyridine-5-boronic acid, 6-(Morpholino)pyridine-3-boronic acid, 6-Morpholinopyridine-3-boronic acid, 6-Morpholino-3-pyridineboronic Acid
BB-8632-5G
BB-8632-1G
COBBBB-8632-5GEA
0
EUR
COBBBB-8632-5G
COBBBB-8632-1G
2-Morpholino-5-pyridinboronsäure
2-Morpholino-5-pyridinboronsäure
Formel:
C₉H₁₃BN₂O₃ |
MDL:
MFCD03086116 CAS-Nummer: 904326-93-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...