3,5-Bis(trifluormethyl)benzylchlorid
Lieferant: FLUOROCHEM
Synonyme:
1-(Chloromethyl)-3,5-bis(trifluoromethyl)benzene
006024-5G
006024-25G
FLUO006024-5GEA
0
EUR
FLUO006024-5G
FLUO006024-25G
3,5-Bis(trifluormethyl)benzylchlorid
3,5-Bis(trifluormethyl)benzylchlorid
Formel:
C₉H₅ClF₆ Molecular Weight: 262.58 g/mol |
MDL:
MFCD00009906 CAS-Nummer: 75462-59-8 EINECS: 278-216-1 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...