2-Methoxy-5-nitro-3-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Methoxy-5-nitro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

PN-5572-1G PN-5572-250M
COBBPN-5572-1GEA 967 EUR
COBBPN-5572-1G COBBPN-5572-250M
2-Methoxy-5-nitro-3-pyridinboronsäurepinakolester
2-Methoxy-5-nitro-3-pyridinboronsäurepinakolester
Formel: C₁₂H₁₇BN₂O₅
Molecular Weight: 280.0900 g/mol
MDL: MFCD08063082
CAS-Nummer: 1218791-18-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR