2-Ethoxy-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Ethoxypyridin-3-yl)boronic acid

BB-2271-5G BB-2271-1G
COBBBB-2271-5GEA 193 EUR
COBBBB-2271-5G COBBBB-2271-1G
2-Ethoxy-3-pyridinboronsäure
2-Ethoxy-3-pyridineboronic acid
Formel: C₇H₁₀BNO₃
Molecular Weight: 166.97 g/mol
MDL: MFCD06201036
CAS-Nummer: 854373-97-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR