2-Methoxy-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

BB-3880-5G BB-3880-1G
COBBBB-3880-5GEA 0 EUR
COBBBB-3880-5G COBBBB-3880-1G
2-Methoxy-4-pyridinboronsäure
2-Methoxy-4-pyridineboronic acid
Formel: C₆H₈BNO₃
MDL: MFCD07368877
CAS-Nummer: 762262-09-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR