2-Ethoxy-5-pyridinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Ethoxy-5-pyridylboronic acid, 6-Ethoxy-3-pyridylboronic acid, 2-Ethoxypyridine-5-boronic acid, (6-Ethoxypyridin-3-yl)boronic acid, 6-Ethoxypyridine-3-boronic acid
BB-2278-1G
BB-2278-25G
BB-2278-5G
COBBBB-2278-1GEA
38.5
EUR
COBBBB-2278-1G
COBBBB-2278-25G
COBBBB-2278-5G
2-Ethoxy-5-pyridinboronsäure
2-Ethoxy-5-pyridinboronsäure
Formel:
C₇H₁₀BNO₃ Molecular Weight: 166.97 g/mol |
MDL:
MFCD06201032 CAS-Nummer: 612845-44-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...