2-Methoxy-5-pyridinboronsäure

Lieferant: CHEMPUR

Synonyme: (6-Methoxypyridin-3-yl)boronic acid

AC35955.0.1 AC35955.250
CHMPAC35955.0.1EA 0 EUR
CHMPAC35955.0.1 CHMPAC35955.250
2-Methoxy-5-pyridinboronsäure
2-Methoxy-5-pyridinboronsäure
Formel: C₆H₈BNO₃
MDL: MFCD02093044
CAS-Nummer: 163105-89-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR